Få mere at vide
Copper(II) Acetate Monohydrate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Copper(II) Acetate Monohydrate |
| CAS | 463931-62-6 |
| Molekylær formel | C4H8O4 .Cu .H2O |
| Mængde | 250 g |
| Synonym | Acetic Acid Copper(2+) Salt Monohydrate, Copper Acetate Monohydrate, Acetic Acid Cupric Salt Monohydrate, Copper Diacetate Monohydrate, Copper(2+) Acetate Monohydrate, Copper(2+) Diacetate Monohydrate, Crystallized Verdigris Monohydrate, Crystals of Venus Monohydrate, Cupric Acetate Monohydrate, Cupric Diacetate Monohydrate, Neutral Verdigris Monohydrate, Ploss Blue Monohydrate, Venus Copper Monohydrate |
| InChI formel | InChI=1S/2C2H4O2.Cu/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2 |
| SMIL | [Cu+2].CC(=O)[O-].CC(=O)[O-] |
| IUPAC navn | copper;diacetate |
| Molekylvægt (g/mol) | 181.63 |