missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) trifluoromethanesulfonate benzene complex, 90%, ACROS Organics™

297.00 DKK - 924.00 DKK

Chemical Identifiers

CAS 42152-46-5
Molecular Formula C8H6Cu2F6O6S2
Molecular Weight (g/mol) 503.33
MDL Number MFCD00043300
Synonym Trifluoromethanesulfonic acid, copper(I) salt, benzene complex
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10028971
SDS View Documents
Acros Organics
371090010
1g Glass bottle
297.00 DKK
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 5 Please to purchase this item. Need a web account? Register with us today!
10293052
SDS View Documents
Acros Organics
371090050
5g Glass bottle
924.00 DKK
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

42152-46-5
503.33
Trifluoromethanesulfonic acid, copper(I) salt, benzene complex
C8H6Cu2F6O6S2
MFCD00043300

Specifications

Copper(I) trifluoromethanesulfonate benzene complex
Authentic
Gray-Green
1g
90%
(CF3SO3Cu)2C6H6
12,144; 16,99
Solubility in water: .. Other solubilities: soluble in benzene,toluene
503.33
90%
7°C
Glass bottle
160.0°C
42152-46-5
C8H6Cu2F6O6S2
MFCD00043300
Trifluoromethanesulfonic acid, copper(I) salt, benzene complex
503.33
Powder
Safety and Handling

Safety and Handling

Product Identifier
  • Copper(I) trifluoromethanesulfonate benzene complex

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS