missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) thiophene-2-carboxylate, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Reactant or reagent used for studies of xenobiotic response safener derivatives, synthesis of functionalized BODIPY dye analogs, orthogonal cross-coupling reactions, preparation of parent borondipyrromethene system and Copper mediated cross-coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant or reagent used for studies of xenobiotic response safener derivatives, synthesis of functionalized BODIPY dye analogs, orthogonal cross-coupling reactions, preparation of parent borondipyrromethene system and Copper mediated cross-coupling.
Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air. Keep container tightly closed. Store away from oxidizing agent.
Reactant or reagent used for studies of xenobiotic response safener derivatives, synthesis of functionalized BODIPY dye analogs, orthogonal cross-coupling reactions, preparation of parent borondipyrromethene system and Copper mediated cross-coupling.
Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Color | Brown to Red |
Physical Form | Powder |
Quantity | 1 g |
Formula Weight | 190.68 |
Sensitivity | Air and light sensitive |
Chemical Name or Material | Copper(I) thiophene-2-carboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'