missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) Oxide, Honeywell Fluka™
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Color | Red |
Quantity | 500 g |
UN Number | 3077 |
Formula Weight | 143.09g/mol |
Percent Purity | ≥86% (Cu basis) |
Packaging | Plastic Bottle |
Chemical Name or Material | Copper(I) Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'