Få mere at vide
Copper Hexafluorosilicate Hydrate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Copper(II) Hexafluorosilicate Monohydrate |
| CAS | 2123555-14-4 |
| Molekylær formel | Cu . F6 Si . H2 O |
| Mængde | 5 g |
| Synonym | Copper Fluosilicate Hydrate, copper(2+) Hexafluorosilicate(2-) Hydrate, Copper Fluorosilicate Hydrate, Copper Fluorosilicate Hydrate, Copper Hexafluorosilicate Hydrate, Copper(2+) Hexafluorosilicate(2-) Hydrate, Cupric Fluorosilicate Hydrate, Cupric Hexafluorosilicate Hydrate, Cupric Silicofluoride Hydrate |
| InChI formel | InChI=1S/Cu.F6Si.H2O/c;1-7(2,3,4,5)6;/h;;1H2/q+2;-2; |
| SMIL | O.[Cu+2].F[Si-2](F)(F)(F)(F)F |
| IUPAC navn | copper;hexafluorosilicon(2-);hydrate |
| Molekylvægt (g/mol) | 223.637 |
| Formel vægt | 222.908 |