missing translation for 'onlineSavingsMsg'
Learn More
Copper, Leaded Tin Bronze Navy M , CDA 922, C92200, ARMI∣MBH Product Code.: 18003330

Copper, Leaded Tin Bronze Navy M , CDA 922, C92200, ARMI∣MBH

Product Code. 18003330
Each
Click to view available options
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 18003330

Brand: ARMI/MBH IARM90CG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Copper, Leaded Tin Bronze Navy M , CDA 922, C92200, Ø38x19mm

Specifications

Chemical Name or Material Cadmium/Chromium/Cobalt/Silicon/Selenium/Arsenic/Manganese/Bismuth/Silver/Antimony/Phosphorus/Iron/Nickel/Lead/Zinc/Tin/Copper/Magnesium/Zirconium/Beryllium/Nitrogen/Niobium/Aluminium/Oxygen/Carbon/Tellurium
CAS 7440-50-8
Concentration or Composition (by Analyte or Components) Cadmium 0.0003 %/Chromium 0.001 %/Cobalt 0.0012 %/Silicon 0.0012 %/Selenium 0.0013 %/Arsenic 0.002 %/Manganese 0.003 %/Bismuth 0.009 %/Silver 0.011 %/Antimony 0.022 %/Phosphorus 0.026 %/Iron 0.101 %/Nickel 0.266 %/Lead 1.4 %/Zinc 4.3 %/Tin 6.3 %/Copper 87.5 %/Magnesium (0.0002) %/Zirconium (0.0002) %/Beryllium (0.0003) %/Nitrogen (0.0003) %/Niobium (0.0005) %/Aluminium (0.001) %/Oxygen (0.001) %/Carbon (0.002) %/Tellurium (0.003) %
Form Disc
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper, Leaded Tin Bronze Navy M , CDA 922, C92200, ARMI∣MBH >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.