missing translation for 'onlineSavingsMsg'
Learn More

Copper, ACS Reagent, Thermo Scientific™

Catalog Number 13256339
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
Melting Point 1083.0°C
Boiling Point 2595.0°C
Linear Formula Cu
Quantity 500 g
Formula Weight 63.54
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Copper

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'