Learn More
Epredia™ Cool-Cut with universal cassette clamp
Maintain a cool, consistent temperature for paraffin blocks during the sectioning process with the Epredia™ Cool-Cut accessory.
35920.00 DKK
Specifications
Product Type | Microtome Accessory |
---|---|
For Use With (Equipment) | Epredia Microtomes |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12617886
|
Epredia™
771120 |
With standard clamp |
35920.00 DKK
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Directly fits onto the specimen cylinder of the rotary microtome line
- Ability to be equipped as a universal cassette clamp or standard specimen clamp
- Effective when extended cutting periods on the microtome are required to “step section” or “serial section” through the paraffin block
Includes:
- Peltier device with integrated specimen clamp
- Counter-balance weights
- Power supply
Specifications
Microtome Accessory | |
Epredia Microtomes |