Learn More
HUBER USB Control Cable
Product Code. 15286537
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
15286537 1 Ea. Each
1 options
Product Code. 15286537 Supplier HUBER Supplier No. 54949
  • ONLINE EXCLUSIVE PRICE
    129.00 DKK
    113.69 DKK / Pakke med 1
    Spare 15.31 DKK
    11% Slukket
Forventet forsendelse: 22-01-2027
USB control cable from: controller Pilot ONE, Mini-USB, to: PC / Notebook, USB Type A. For data recording, data communication and firmware updates.
Specifications
Product Type USB Control Cable
Quantity 1 Ea.