Learn More
Macherey-Nagel™ Congo paper MN 816 N pH Paper
Brand: Macherey-Nagel™ 90702
Additional Details : Weight : 0.02070kg
Specifications
Red to Blue | |
Reel | |
1 Each | |
pH Paper |
5 to 3 | |
Determination of lactic acid | |
Determination of lactic acid | |
7 mm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Congo paper
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : Congo red