Learn More
Conductivity standard solution 350,000μS/cm conductivity at 25°C traceable to NIST, Reagecon™
BIOPHARMA
Brand:  Reagecon CSKC350M
Quantity 500 mL
Product Code. 11596723
  • 1210.00 DKK / 500mL
Specifications
500 mL
Documents
Brochures
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCS350M24L1 Certificate TypeCertificate of Analysis Date16/04/2025 Catalog NumberCSKC350M
Lot NumberCS350M23M1 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog NumberCSKC350M
Lot NumberCS350M22L1 Certificate TypeCertificate of Analysis Date23/05/2023 Catalog NumberCSKC350M
Lot NumberCS350M22L1 (1) Certificate TypeCertificate of Analysis Date23/05/2023 Catalog NumberCSKC350M
Lot NumberCS350M21M2 Certificate TypeCertificate of Analysis Date06/04/2022 Catalog NumberCSKC350M
Promotions
Promotions
Product Content Correction
Product Title
Conductivity standard solution 350,000μS/cm conductivity at 25°C traceable to NIST, Reagecon™ >