Få mere at vide
Scharlab Conductivity standard, 1413 µS/cm (25 °C), KCl0,01 mol/l, 250 mL, MA: PA01010250, MSDS:
Brand:  Scharlab PA01010250
Conductivity standard, 1413 µS/cm (25 °C), KCl0,01 mol/l, 250 mL, MA: PA01010250, MSDS:
Produktkode 12477312
  • ONLINE EXCLUSIVE PRICE
    1806.00 DKK
    193.49 DKK / 250ml
    Save 1612.51 DKK
    89% Slukket