Få mere at vide
Jenway™ 3540 Bench Combined Conductivity/pH Meter
Artikelnummer. 11400609
:
Model: 3540
Artikelnummer. 11400609
Brand: Jenway™ 1008828
Designed for use in all laboratories where pH and conductivity analyses are required. Jenway™ 3540 Bench Combined Conductivity/pH Meters feature automatic calibration, and whole range of instrument, pH and conductivity measurement options.
  • Simultaneous display of pH and conductivity
  • 2 independent channels to prevent interference between probes
  • Additional modes for resistivity, salinity, TDS and mV
  • Data logger with auto save/print options
  • Storage of up to 500 readings (250 for each mode)