Learn More
3M™ Comply™ SteriGage™ Steam Chemical Indicator
Product Code. 11943604
Change view
Click to view available options
Quantity:
500 Pk.
Unit Size:
Pack of 500
Product Code. Quantity Unit Size
11943604 500 Pk. Pack of 500
1 options
Product Code. 11943604 Supplier 3M™ Supplier No. 7000002707
  • ONLINE EXCLUSIVE PRICE
    2868.31 DKK
    1070.94 DKK / Pakke med 500
    Spare 1797.37 DKK
    62% Slukket
Forventet forsendelse: 22-12-2026
Offers immediate, accurate and easy to read method of monitoring the sterilization process conditions inside each pack and can be used in all steam sterilizers. 3M™ Comply™ SteriGage™ Steam Chemical Indicator with easy-to-read moving front style has been shown to have a higher reading accuracy than color interpretation style indicators.
Offers immediate, accurate and easy to read method of monitoring the sterilization process conditions inside each pack and can be used in all steam sterilizers. Easy-to-read moving front style has been shown to have a higher reading accuracy than color interpretation style indicators.
  • Moving front ink technology provides easy to read 'Accept or Reject' result
  • Can be used in all 121°C to 135°C steam sterilization cycles
  • Response correlates with a Biological Indicator at 3 time/temperature relationships under ideal steam sterilization conditions.
  • Confirmed by BSI, a leading global independent Product Testing Service Company, to meet the performance requirements of ANSI/AAMI/ISO 11140-1:2005 Class 5
Specifications
Certifications/Compliance ISO11140
For Use With (Application) Determining quality of sterilization
For Use With (Equipment) Sterilizer
Product Type Steam Chemical Indicator
Temperature (Metric) 121°C
Quantity 500 Pk.
Sterilization Indicator Yes
Sterilization Method Steam, Chemical