Få mere at vide
Lovibond™ Comparator 2000+ disc for Hazen scale NSH
Beskrivelse
Comparator 2000+ disc for Hazen scale NSH range 10-20-30-40-50-60-70-80-90 Pt-Co for use with Nessleriser 2150
Resistance
The color standards made of glass are resistant to light and chemicals. This ensures a long life of the color disc.
Shock resistant
Thanks to its high hardness and scratch resistance, the color disc is resistant to case damage.
Colour stable
Lovibond™ color standards consist exclusively of through-and-through colored and flat-ground glasses.
Tekniske data
Tekniske data
| Rækkevidde | 10 to 20 to 30 to 40 to 50 to 60 to 70 to 80 to 90 Pt-Co |
| Type | Hazen Scale NSH Comparator Disc |