Learn More
Nabertherm™ Compact Tube Furnace with C550 Controller
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
Brand:  Nabertherm™ R-252O3AN
 View more versions of this product
Product Code. 17388079
  • 45355.00 DKK / Each
Only null left
Estimated Shipment: 22-07-2025
Description
  • Excellent price - performance ratio
  • Can also be equipped with gas supply system 1 (no vacuum operation) and gas supply system 2 (for vacuum operation)
  • Temperature, max. 1200°C or 1300°C
  • Five programs and four freely programmable ramps, controller C 550 with 10 freely adjustable programs and 20 segments
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Dual shell housing made of sheets of textured stainless steel
  • Insulation made of non-classified fibre material
Specifications
Specifications
C550
1300°C
670 x 340 x 508 mm
50 mm
500 mm
Compact Tube Furnace
700 mm
Documents
Brochures
Product Guides
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Nabertherm™ Compact Tube Furnace with C550 Controller > Diameter: 50 mm; Heated Zone: 500 mm; Max Temp: 1300°C