Learn More
Nabertherm™ Compact Tube Furnace
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
Brand: Nabertherm™ R-151C3AN
Additional Details : Weight : 22.00000kg
Description
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Solid state relays provide for low-noise operation
Specifications
C450 | |
1300°C | |
450 mm |
250 mm | |
434 x 340 x 508 mm | |
50 mm |