missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMMD9 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-00330AF647
Additional Details : Weight : 0.00970kg
Description
COMMD9 Polyclonal specifically detects COMMD9 in Human samples. It is validated for ELISA.Specifications
COMMD9 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
29099 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 647 | |
COMM domain containing 9, COMM domain-containing protein 9, COMM9, FLJ31106, HSPC166 | |
Produced in rabbits immunized with purified, recombinant COMMD9 (Q9P000-1; Met1-Lys198). | |
0.1 mL | |
Primary | |
Human | |
Purified |