Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1.5mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11887583
  • 153.00 DKK / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Recommended for maximising sample volume
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Mini SDS PAGE, Native PAGE, Gradient, Second Dimension and Nucleic Acid Separations
Comb
1 Each
60 μL
OmniPAGE Mini Vertical Gel Electrophoresis Unit
12
1.5 mm