Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 1mm Thick
Product Code. 11893373
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
14
16
18
2 Prep, 2 Marker
20
30
35
4 Prep, 2 Marker
Volume (Metric) Sample:
10 μL
11 μL
13 μL
21 μL
27 μL
29 μL
41 μL
45 μL
122 μL
225 μL
495 μL
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Sample Unit Size
11893373 14 29 μL Each
11843363 10 45 μL Each
11883363 10 29 μL Each
11813373 1 Prep, 1 Marker 495 μL Each
11853373 12 41 μL Each
11833383 16 27 μL Each
11873383 18 11 μL Each
11823393 20 21 μL Each
11853393 2 Prep, 2 Marker 225 μL Each
11833403 30 13 μL Each
11873403 35 10 μL Each
11813413 4 Prep, 2 Marker 122 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 11893373
  • ONLINE EXCLUSIVE PRICE
    219.00 DKK
    209.63 DKK / Pakke med 1
    Spare 9.37 DKK
    4% Slukket
Forventet forsendelse: 21-12-2026
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • Supplied as standard
  • Recommended for high resolution gels and publication quality data
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
For Use With (Equipment) MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 14
Quantity 1 Each
Thickness (Metric) 1 mm
Volume (Metric) Sample 29 μL