Learn More
Fisherbrand™ Combs for MultiSUB Maxi Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11823433
  • 261.00 DKK / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • Recommended for high resolution gels and publication quality data
  • Supplied as standard
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
Specifications
MultiSUB Maxi-Gel Electrophoresis Unit
16
1 mm
Comb
1 Each
41 μL