Learn More
Hoefer™ Combs for HE 33 Mini Submarine Unit
Accessory parts for HE 33 Mini Submarine
Brand: Hoefer™ HE31A-8-1.5
Additional Details : Weight : 0.45360kg
Description
- Variety of eight sizes ranging from 1/2 to 16 wells
- 1 to 1.5 mm Thickness
- 44/6 to 2.6 mm Well Width
- 235/35 to 20.5 μL Sample Volume
Specifications
HE33 Mini Submarine Unit | |
8 | |
0.15 cm | |
0.25 in. | |
0.19 in. |
Backless Comb | |
0.05 in. | |
48.5 μL | |
0.65 cm | |
0.5 cm |