missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 0.75mm Thick

For use in preparatory electrophoresis and high-throughput screening.

Brand:  Fisherbrand™

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11897603

  • 141.00 DKK / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Recommended for ultra resolved bands
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications

Specifications

Black
OmniPAGE Mini Vertical Gel Electrophoresis Unit
5
0.75 mm
Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations
Comb
1 Each
70 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers