Få mere at vide
Combretastatin A4, TRC
Artikelnummer. 30356464
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356464 50 mg
30367660 5 mg
30357201 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30356464 Leverandør Toronto Research Chemicals Leverandørnr. TRCC64300050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 117048-59-6
Molekylær formel C18 H20 O5
Molekylvægt (g/mol) 316.35
IUPAC navn 2-methoxy-5-[(Z)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenol
SMIL COc1ccc(\C=C/c2cc(OC)c(OC)c(OC)c2)cc1O
Tekniske data
Kemisk navn eller materiale Combretastatin A-4
Mængde 50 mg
InChI formel InChI=1S/C18H20O5/c1-20-15-8-7-12(9-14(15)19)5-6-13-10-16(21-2)18(23-4)17(11-13)22-3/h5-11,19H,1-4H3/b6-5-
Formel vægt 316.1311