Få mere at vide
WTW™ Combined Oxygen-/Conductivity Sensors
Description
- Combined Oxygen/Conductivity sensor
- Includes a cable 9.8 ft (3m) and instructions for use, h 3 exchange membrane heads, cleaning solution, conductivity standard (1413μS/cm), abrasive foil, OxiCal® CX calibration vessel, electrolyte
- Combined Oxygen-/Conductivity sensor with cable 9.8 ft (3 m) and instructions for use
- With 3 exchange membrane heads, cleaning solution, conductivity standard (1413 μS/cm), abrasive foil, OxiCal CX calibration vessel, electrolyte
- Measures oxygen, conductivity and temperature simultaneously
Specifications
Specifications
| Oversigt | Combined Oxygen/Conductivity sensor |
| Inkluderer | Cable 9.8 ft (3m) and instructions for use, h 3 exchange membrane heads, cleaning solution, conductivity standard (1413μS/cm), abrasive foil, OxiCal™ CX calibration vessel, electrolyte |
| Længde (metrisk) | 3 m |