Learn More
Fisherbrand™ Combs for TV200 and TV400 Twin-Plate Vertical Gel Electrophoresis Unit
For use with TV200 Standard Twin-Plate Wide Format Mini-Gel and TV400 Standard Twin-Plate Maxi-Gel vertical electrophoresis units
Brand:  Fisherbrand™
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 10448212
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Available in 0.75, 1, 1.5 and 2mm thickness
Specifications
Specifications
24
8 μL
Teeth: 5 mm
0.35 mm
4.75 mm