Learn More
Fisherbrand™ Combs for omniPAGE Mini Vertical Gel Electrophoresis Unit, 0.5mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- 0.5mm thick
- 330μL sample size
Specifications
Vertical Gel Electrophoresis System | |
1 Prep, 1 Marker | |
330 μL |
Comb | |
0.5 mm |