Learn More
Ohaus™ Column Mount Kit, 700 mm, Stainless Steel for Scale
Suitable stainless steel, washdown-capable Defender series scale column connects base scale with readout and control panel.
Brand: Ohaus™ 80251746
Additional Details : Weight : 2.88000kg
Description
- Stainless steel construction.
- Compatible with Defender series scales.
- Base, D30BR
- Base, D60BR
- Base, D60BL
- Base, D150BL
- Base, D150BX
- Base, D300BX
- Base, D60VR
- Base, D60VL
- Base, D150BX
- Base, D300BX
- Base, D60VR
- Base, D60VL
Specifications
Defender Series Scales | |
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) | |
Column Mounts |
Stainless Steel | |
700 mm |