Learn More
Lauda™ Colorless Heat Transfer Liquid
Brand:  Lauda™ LZB 102
Product Code. 12052532
  • 5180.00 DKK / Each
Estimated Shipment: 08-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
-60°C to +80°C
Kryo 60
3mm2/sec.
Bath Filler
5 L