Learn More
Fisherbrand™ Colored PTFE Conical Joint Clamp
Fisherbrand™ Colored PTFE Conical Joint Clamps for glass joints and can be used as a safe solution to clamping glass joints. The PTFE conical joint clamps include an encapsulated steel spring.
Brand:  Fisherbrand™
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 16286041
  • 317.00 DKK / Pack of 5
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
  • Chemically inert and chemically resistant
  • Temperature resistant, maintaining strength up to 280°C
  • Fit stndard glass joints
  • Can be adjusted by hand
NOTE: Colored PTFE is not as inert as pure PTFE
Specifications
Specifications
1
Standard Taper Ground Glass Joints
Joint 45/50
PTFE
Fisherbrand™ Colored PTFE Conical Joint Clamp
Brown