missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Colorectal Cancer Associated 1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-05947AF700
Additional Details : Weight : 0.00970kg
Description
Colorectal Cancer Associated 1 Polyclonal specifically detects Colorectal Cancer Associated 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Colorectal Cancer Associated 1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C11orf92, chromosome 11 open reading frame 92, DKFZp781P19126, FLJ45803 | |
Produced in rabbits immunized with E. coli-derived Human Colorectal Cancer Associated 1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
399948 | |
Store at 4°C in the dark. | |
IgG |