Få mere at vide
Collidine-hydrogen Fluoride Complex (approx 1:2 collidine:HF), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Collidine-hydrogen Fluoride Complex (1:2 collidine:HF) |
| CAS | 45725-47-1 |
| Molekylær formel | C8H11N . 2HF |
| Mængde | 250 mg |
| Synonym | Pyridine, 2,4,6-trimethyl-, hydrofluoride (1:1) (ACI), Pyridine, 2,4,6-trimethyl-, hydrofluoride (9CI), 2,4,6-Trimethylpyridine hydrofluoride, Collidine-hydrogen Fluoride Complex |
| InChI formel | InChI=1S/C8H11N.FH/c1-6-4-7(2)9-8(3)5-6;/h4-5H,1-3H3;1H |
| SMIL | F.Cc1cc(C)nc(C)c1 |
| IUPAC navn | 2,4,6-trimethylpyridine;hydrofluoride |
| Molekylvægt (g/mol) | 141.19 |
| Formel vægt | 141.0954 |