Beskrivelse
The collection chamber is available in two sizes, suited to the respective volume range of the tips. It features a smooth surface that can be cleaned without residue and sterilised. The collection chamber ensures that the collected powder sample is securely stored until it is analysed and enables rapid volume changes by simply exchanging the tips.
- Stainless steel collection cylinder for PharmaPicker - container for the collected sample
Tekniske data
Tekniske data
| Produkttype | Sampler Insert |
| Materiale | Stainless Steel (V4A) |
| Autoklaverbar | Autoclavable |
| Kapacitet (metrisk) | 0.1 to 1.2 mL |