Learn More
Collagenase, Type I from Clostridium histolyticum, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
1 g
5 g
Description
- Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
Specifications
Concentration | ≥125 U/mg |
Components | Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes |
Content And Storage | Store desiccated at 4°C |
Source | Clostridium histolyticum |
Quantity | 100 mg |
Product Type | Collagenase |
Form | Lyophilized |