missing translation for 'onlineSavingsMsg'
Learn More

Collagenase, Type I from Clostridium histolyticum, Thermo Scientific Chemicals

Catalog Number 15875448
Click to view available options
Quantity:
100 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes

  • Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
TRUSTED_SUSTAINABILITY
Concentration ≥125 U/mg
Components Contains mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes
Content And Storage Store desiccated at 4°C
Source Clostridium histolyticum
Quantity 100 mg
Product Type Collagenase
Form Lyophilized

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'