missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen I Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-408PE
This item is not returnable.
View return policy
Description
Collagen I Polyclonal antibody specifically detects Collagen I in Human, Mouse, Rat, Porcine, Bovine, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), SDS-Page, Fluorophore-linked immunosorbent assay, Microarray, KnockDown
Specifications
Collagen I | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
PE | |
Alpha-1 type I collagen, alpha1(I) procollagen, CAFYD, COL1A1, Collagen 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen alpha-1(I) chain preproprotein, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, EDSARTH1, EDSC, OI1, OI2, OI3, OI4, pro-alpha-1 collagen type 1, type I proalpha 1, Type I Procollagen Alpha 1 Chain | |
Collagen I from human and bovine placenta (Uniprot: P02452) | |
0.1 mL | |
Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Stem Cells | |
1277 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Collagen I Antibody, PE, Novus Biologicals™ > 0.1 mL; PE
Spot an opportunity for improvement?Share a Content Correction