Få mere at vide
Colchicine-d6, TRC
Artikelnummer. 30367538
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30367538 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367538 Leverandør Toronto Research Chemicals Leverandørnr. TRCC64000310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Colchicine-d6
CAS 1217651-73-4
Renhedsgrad noter HPLC
Molekylær formel C22 H19 D6 N O6
Mængde 10 mg
Synonym 2,2,2-Trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[d]heptalen-7-yl]acetamide, Colchicine-d6, Colchicine-D6 (7-acetamide-D3,10-methoxy-D3), N-[(7S)-1,2,3-Trimethoxy-10-(methoxy-d3)-9-oxo-5,6,7,9-tetrahydrobenzo[a]heptalen-7-yl]acetamide-2,2,2-d3, N-[(7S)-5,6,7,9-Tetrahydro-1,2,3-trimethoxy-10-(methoxy-d3)-9-oxobenzo[a]heptalen-7-yl]acetamide-2,2,2-d3
InChI formel InChI=1S/C22H25NO6/c1-12(24)23-16-8-6-13-10-19(27-3)21(28-4)22(29-5)20(13)14-7-9-18(26-2)17(25)11-15(14)16/h7,9-11,16H,6,8H2,1-5H3,(H,23,24)/t16-/m0/s1/i1D3,2D3
SMIL [2H]C([2H])([2H])OC1=CC=C2C(=CC1=O)[C@H](CCc3cc(OC)c(OC)c(OC)c23)NC(=O)C([2H])([2H])[2H]
IUPAC navn 2,2,2-trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
Molekylvægt (g/mol) 405.472
Formel vægt 405.2058
Procent renhed >95