Learn More
Abnova™ COL1A1 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
12030.00 DKK - 19810.00 DKK
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Format | Tube |
For Use With (Application) | FISH-Ce |
Gene ID (Entrez) | 1277 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063475
|
Abnova™
FS0043.100uL |
100 ÎĽL |
12030.00 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16073475
|
Abnova™
FS0043.200uL |
200 ÎĽL |
19810.00 DKK
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Store at 4°C in the dark. | |
Tube | |
1277 | |
Liquid |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Abnova™ COL1A1 Split FISH Probe