missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
COG6 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18428JF646
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
COG6 Polyclonal antibody specifically detects COG6 in Human, Mouse samples. It is validated for Western Blot
Tekniske data
| COG6 | |
| Polyclonal | |
| Western Blot | |
| COG complex subunit 6, component of oligomeric golgi complex 6KIAA1134COD2complexed with Dor1p 2, conserved oligomeric Golgi complex protein 6, conserved oligomeric Golgi complex subunit 6, DKFZp313D191 | |
| Recognizes region between residues Between Between 607 and 657.. | |
| 0.1 mL | |
| Golgi Apparatus Markers | |
| 57511 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion