missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme A Trilithium Salt Dihydrate, Thermo Scientific Chemicals

Catalog Number 15855268
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
18439-24-2
C21H33Li3N7O16P3S
785.329
MFCD00075848
QSCBPHBAFBVXRK-UHFFFAOYSA-K
coenzyme a, trilithium salt
53442190
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
This item is not returnable. View return policy
18439-24-2
C21H33Li3N7O16P3S
785.329
MFCD00075848
QSCBPHBAFBVXRK-UHFFFAOYSA-K
coenzyme a, trilithium salt
53442190
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
Packaging Plastic bottle
Quantity 250 mg
Formula Weight 821.36
Physical Form Crystalline Powder
Chemical Name or Material Coenzyme A trilithium salt dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'