Få mere at vide
Codeinone, Mikromol™
Beskrivelse
Codeinone
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Codeinone |
| CAS | 467-13-0 |
| Analyt- eller komponentnavne | Codeinone |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H19 N O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Hydrocodone Hydrogen Tartrate 2.5-Hydrate Imp. E (EP), Hydrocodone Imp. E (EP), Oxycodone USP Related Compound C CII, Oxycodone USP RC C CII, 7,8-Didehydro-4,5alpha-epoxy-3-methoxy-17-methylmorphinan-6-one, Codeinone, Oxycodone Related Compound C, Codeine Monohydrate Impurity I, Hydrocodone Hydrogen Tartrate 2.5-Hydrate Impurity E, Codeine Impurity I, Hydrocodone Impurity E |
| InChI formel | InChI=1S/C18H19NO3/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19/h3-6,11-12,17H,7-9H2,1-2H3/t11-,12+,17-,18-/m0/s1 |
| SMIL | COc1ccc2C[C@@H]3[C@@H]4C=CC(=O)[C@@H]5Oc1c2[C@]45CCN3C |
| IUPAC navn | (4R,4aR,7aR,12bS)-9-methoxy-3-methyl-1,2,4,4a,7a,13-hexahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one |
| Vis mere |