Få mere at vide
Coconut Oil Monoethanolamide, TRC
Artikelnummer. 30179649
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179649 50 mg
30179557 10 mg
30179091 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179649 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18205550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 142-54-1
Molekylær formel C15H31NO2
Molekylvægt (g/mol) 257.41
Synonym N-(2-Hydroxypropyl)dodecanamide, 2-Hydroxypropyllauramide, Alkamide LIPA, Amisol PLME, Amisol PLME-A, Amizett PLME-A, Clindrol 101LI, Clindrol 102LI, Comperlan LP, Cyclomide LP, LIPA, LPA, Lauramide MIPA, Lauric acid isopropanolamide, Lauric acid monoisopropanolamide, Lauric isopropanolamideLauric monoisopropanolamide, Lauroyl isopropanolamide, Lauryl isopropanolamideLauryl monoisopropanolamide, N-(2-Hydroxy-1-propyl)lauramide, N-(beta-Hydroxypropyl)lauramide, Profan AD31, Stafoam LIPA, Steinamid IPL 203, Ultrapole L
IUPAC navn N-(2-hydroxypropyl)dodecanamide
SMIL CCCCCCCCCCCC(=O)NCC(C)O
Tekniske data
Kemisk navn eller materiale Coconut Oil Monoethanolamide
Mængde 50 mg
InChI formel InChI=1S/C15H31NO2/c1-3-4-5-6-7-8-9-10-11-12-15(18)16-13-14(2)17/h14,17H,3-13H2,1-2H3,(H,16,18)
Formel vægt 257.2355