Få mere at vide
Cobimetinib hemifumarate, MedChemExpress
Artikelnummer. 16485528
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16485528 10 mg 10mg
16401707 50 mg -
16478237 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16485528 Leverandør Medchem Express Leverandørnr. HY13064A
MedChemExpress Cobimetinib hemifumarate is a novel selective MEK1 inhibitor, and the IC50 value against MEK1 is 4.2 nM.
Cobimetinib hemifumarate is a novel selective MEK1 inhibitor, and the IC50 value against MEK1 is 4.2 nM.
Tekniske data
Kemisk navn eller materiale Cobimetinib hemifumarate
CAS 1369665-02-0
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C21H21F3IN3O2.1/2C4H4O4
Mængde 10 mg
Synonym GDC-0973 hemifumarateXL-518 hemifumarate
Opløselighedsinformation DMSO : 50 mg/mL (84.85 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.70 mM; ultrasonic and warming and heat to 60°C)
SMIL OC1(CN(C(C2=C(NC3=C(F)C=C(I)C=C3)C(F)=C(F)C=C2)=O)C1)[C@H]4NCCCC4.O=C(/C=C/C(O)=O)O.[0.5]
Molekylvægt (g/mol) 589.25
Formel vægt 589.25
Procent renhed 96.76%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease