missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) stearate, Co 9-10%, Thermo Scientific™

Catalog Number p-7058835
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
20 kg
This item is not returnable. View return policy
1002-88-6
C36H70CoO4
625.89
MFCD00054343
AMFIJXSMYBKJQV-UHFFFAOYSA-L
cobalt ii stearate, cobaltous stearate, cobalt stearate, stearic acid cobalt ii salt, octadecanoic acid, cobalt salt, cobalt distearate, cobalt 2+ dioctadecanoate, stearic acid cobalt salt, cobalt 2+ ; octadecanoate
6451168
cobalt(2+);octadecanoate
[Co++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
This item is not returnable. View return policy
1002-88-6
C36H70CoO4
625.89
MFCD00054343
AMFIJXSMYBKJQV-UHFFFAOYSA-L
cobalt ii stearate, cobaltous stearate, cobalt stearate, stearic acid cobalt ii salt, octadecanoic acid, cobalt salt, cobalt distearate, cobalt 2+ dioctadecanoate, stearic acid cobalt salt, cobalt 2+ ; octadecanoate
6451168
cobalt(2+);octadecanoate
[Co++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Purple
Density 1.71 g/mL
Linear Formula (C18H35O2)2Co
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 625.89
Concentration or Composition (by Analyte or Components) Co 9 to 10%
Physical Form Pellets
Chemical Name or Material Cobalt (II) stearate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'