missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cobalt(II) oxide, 99.998% (metals basis)

Brand:  Alfa Aesar™ 045907.14

Additional Details : CAS Number : 1307-96-6 Weight : 0.02500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15402557

  • 4442.99 DKK / 25g


Estimated Shipment date
from Supplier 19-11-2018
Add to basket

Specifications

Chemical Identifiers

1307-96-6
74.932
IVMYJDGYRUAWML-UHFFFAOYSA-N
oxocobalt
CoO
MFCD00016031
14786
O=[Co]

Specifications

Cobalt (II) oxide
99.998%
CoO
UN2811
IVMYJDGYRUAWML-UHFFFAOYSA-N
oxocobalt
14786
Crystals
6.45g/mL
Air Sensitive
1935°C
1307-96-6
(metals basis)
MFCD00016031
14,2446
O=[Co]
74.932
74.93
CoO, may contain up to 10% Co3O4
3 to 7mm
Brown-Green
25g
Videos
SDS