missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TOKU-E™ Co-trimoxazole ReadyMade™ Solution
Beskrivelse
- CAS Number: 8064-90-2
- Molecular Formula: C14H18N4O3 · 5 C10H11N3O3S
- Molecular Weight: 1556.71 g/mol
- Storage Conditions: -20°C
- Form: Liquid
- Source: Synthetic mixture
- Spectrum: Co-trimoxazole is broad-spectrum, active against Gram-negative bacteria, Gram-positive bacteria, fungi, parasites, and some mycobacteria. Non-susceptible organisms include Mycobacterium tuberculosis, Treponema pallidum, Pseudomonas aeruginosa, and Mycoplasma species
- Mechanism of Action: Co-trimoxazole interferes with bacterial folic acid metabolism by blocking nucleotide biosynthesis. Trimethoprim binds to dihydrofolate reductase (DHFR) which blocks the reduction of dihydrofolic acid (DHF) to tetrahydrofolic acid (THF) and gives the compound its antimicrobial effect. Sulfamethoxazole interferes with nucleic acid synthesis by blocking conversion of p-aminobenzoic acid to the coenzyme dihydrofolic acid. The net result is to inhibit thymine synthesis which in turn prevents the synthesis of bacterial DNA
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Co-Trimoxazole, Cotrimoxazol, Benzenesulfonamide |
| Fysisk form | Liquid |
| CAS | 8064-90-2 |
| Kilde | Synthetic; Mixture |
| Emballage | 2 mL plastic vial |
| Steril filtreret | Yes, 0.22 μm |
| Sterilitet | Sterile |
| Farve | Colorless |
| Koncentration | 100 to 105 mg/mL |
| Mængde | 10 x 1 ml |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?