Få mere at vide
Invitrogen™ CNKSR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113045
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CNKSR3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.
Tekniske data
| CNKSR3 | |
| Polyclonal | |
| Unconjugated | |
| CNKSR3 | |
| 6820402C05; BC024086; CNK homolog protein 3; CNK3; CNKSR family member 3; Cnksr3; Connector enhancer of kinase suppressor of ras 3; connector enhancer of KSR 3; KOX14; MAGI1; maguin-like protein; membrane associated guanylate kinase interacting protein-like 1; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase-interacting protein-like 1; parturition-related protein 4; Prp4; ZNF21 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 154043, 215748 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6P9H4, Q8BMA3 | |
| CNKSR3 | |
| Recombinant Human Connector enhancer of kinase suppressor of ras 3 protein (301-555AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |