Få mere at vide
Electrothermal™ CMUA Series kontrolleret omrørerkappe
Kapacitet (metrisk):
1000 ml
5000 ml
Dimensioner (L x B x H):
220 x 220 x 170 mm
320 x 320 x 220 mm
Watt:
380 W
800 W
Vægt (metrisk) forsendelse:
1,75 kg
2,75 kg
Pakningsstørrelse:
Hver
Beskrivelse
- The electric circuitry for stirring is built into the base of the heater, enabling powerful stirring over a wide range of solution volumes and viscosities using a magnetic stir bar.
- Ideal for busy laboratories with multiple users as they are very safe to use
- Metal case designed to remain cool when in operation.
- Stir speed is bi-directional and easily adjustable
- Auto-recapture switch can be engaged in the event a temporarily interruption in magnetic stirring.
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 5000 ml |
| Dimensioner (L x B x H) | 320 x 320 x 220 mm |
| Spænding | 230 V |
| Elektriske krav | 230v 50/ 60Hz, 800w, With UK and EU plugs |
| Materiale | Powder-coated aluminum |
| Strømforsyningsdetaljer | UK and EU plugs |
| Hastighed | up to 2000rpm |
| Watt | 800 W |
| Vægt (metrisk) forsendelse | 2,75 kg |
| Temperatur (metrisk) | 450°C |
| Vis mere |