Learn More
Gibco™ CMRL Medium, no glutamine
CMRL Medium, no glutamine
Brand: Gibco™ 21530076
5913.62 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
CMRL Medium was originally developed by Connaught Medical Research Laboratories for the growth of Earle's 'L'cells under serum-free conditions. Gibco™ CMRL Medium is also especially useful for cloning monkey kidney cells and for growth of many other mammalian cell lines when supplemented with horse or calf serum.Gibco™ CMRL Medium is a modified version of Medium 199. CMRL medium is particularly rich in nucleosides and vitamins.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.
Dual-Site cGMP Manufacturing
Gibco™ CMRL Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ CMRL Medium product made in our Grand Island facility (11530-037). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
CMRL Medium contains no proteins or growth factors. Therefore, CMRL Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). CMRL Medium uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
CMRL Medium, 500mL, Liquid Form, With Phenol Red, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture | |
CMRL Medium | |
No Glutamine, No HEPES | |
10 x 500 mL |
2 to 8°C | |
Liquid | |
Phenol Red | |
Mammalian cell culture | |
For in-vitro diagnostic use |