Få mere at vide
Tocris Bioscience™ CLP 257
GREENER_CHOICE
Selective KCC2 activator
Brand:  Tocris Bioscience™ 5242/50
Encompass
Produktkode 15786606
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
CLP 257 is a selective KCC2 K+-Cl- cotransporter activator (EC50 = 616 nM). Exhibits selectivity for KCC2 over other KCC family members, NKCC1 and GABAA receptors, as well as 55 other receptors. Enhances Cl- transport and increases KCC2 plasma membrane expression in spinal slices with reduced KCC2 function. Alleviates hypersensitivity in rats with peripheral nerve injury.
Tekniske data
Tekniske data
(5Z)-5-[(4-Fluoro-2-hydroxyphenyl)methylene]-2-(tetrahydro-1-(2H)-pyridazinyl)-4(5H)-thiazolone
1181081-71-9
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner